Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Nucleocapsid (T205I) Protein (His Tag)
Supplier:  Sino Biological 40588V07E9100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Nucleocapsid (YP_009724397.2, with mutation T205I) (Met1-Ala419) was expressed with a polyhistidine tag at the N-terminus. The mutation were identified in the SARS-CoV-2 variant (known as 20C/501Y.V2 or B.1.351 lineage) which emerged in South Africa.
Catalog No. 50-224-6994