Learn More
Sino Biological Anti-CADM1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11168RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CADM1 / IGSF4A extracellular domain (rh CADM1; Catalog#11168-H08H; NP_055148.3; Met 1-His 374). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4812