Learn More
Sino Biological Anti-CADM1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11168RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SynCam / CADM1 (rh SynCam / CADM1; Catalog#11168-H08H; NP_055148.3; Met 1-His 374). SynCam / CADM1 specific IgG was purified by Human SynCam / CADM1 affinity chromatography.
Catalog No. 50-255-4811