Learn More
Sino Biological Recombinant Anti-CADM3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11214R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CADM3 (rh CADM3; Catalog#11214-H08H; NP_001120645.1; Met 1-His 330).
Catalog No. 50-255-4789