Learn More
Sino Biological Anti-CADM3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11214RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CADM3 (rh CADM3; Catalog#11214-H08H; NP_001120645.1; Met 1-His 330). CADM3 specific IgG was purified by human CADM3 affinity chromatography.
Catalog No. 50-255-4787