Learn More
Bachem Calcitonin (human) Trifluoroacetate
SDP
Supplier:  Bachem 4014409.0001
Encompass_Preferred
Calcitonin (human) Trifluoroacetate, H-Cys-Gly-Asn-Leu-Ser-Thr-Cys-Met-Leu-Gly-Thr-Tyr-Thr-Gln-Asp-Phe-Asn-Lys-Phe-His-Thr-Phe-Pro-Gln-Thr-Ala-Ile-Gly-Val-Gly-Ala-Pro-NH2, 21215-62-3, C151H226N40O45S3, Mr 3417.90, 1mg. Calcitonin is a peptide hormone involved in calcium and phosphorus homeostasis. Its inhibitory effect on osteoclast activity lowers blood calcium levels and positively influences bone mass density.
Catalog No. NC1137212