Learn More
Biomatik Corporation Human Calcitonin Gene Related Peptide (CGRP) ELISA Kit
Supplier:  Biomatik Corporation EKL56258
Encompass_Preferred
Human Calcitonin Gene Related Peptide (CGRP) ELISA Kit for detecting CGRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 4.26pg/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 3h. Accession Number: P10092. Gene ID: 797. Target Synonyms: CALCB; CGRP2; CALC2; CGRP-II; Calcitonin Gene Related Peptide 2; Calcitonin 2; Calcitonin Related Polypeptide Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5922