Learn More
Biomatik Corporation Human Calcium/Calmodulin Dependent Protein Kinase II Beta (CAMK2b) ELISA Kit
Supplier:  Biomatik Corporation EKU02871
Encompass_Preferred
Human Calcium/Calmodulin Dependent Protein Kinase II Beta (CAMK2b) ELISA Kit for detecting CAMK2b in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13554. Gene ID: 816. Target Synonyms: CAM2; CAMK2; CAMKB; Proline Rich Calmodulin-Dependent Protein Kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9086