Learn More
Biomatik Corporation Human Calcium/Calmodulin Dependent Protein Kinase II Alpha (CAMK2a) ELISA Kit
Supplier:  Biomatik Corporation EKU02869
Encompass_Preferred
Human Calcium/Calmodulin Dependent Protein Kinase II Alpha (CAMK2a) ELISA Kit for detecting CAMK2a in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UQM7. Gene ID: 815. Target Synonyms: CAMKA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9087