Learn More
Biomatik Corporation Human Calcium/Calmodulin Dependent Protein Kinase Kinase 2 (CAMKK2) ELISA Kit
Supplier:  Biomatik Corporation EKU02873
Encompass_Preferred
Human Calcium/Calmodulin Dependent Protein Kinase Kinase 2 (CAMKK2) ELISA Kit for detecting CAMKK2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96RR4. Gene ID: 10645. Target Synonyms: CAMKK; CAMKKB; Calcium/calmodulin-dependent protein kinase kinase beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5462