Learn More
Biomatik Corporation Human Calcyclin Binding Protein (CACYBP) ELISA Kit
Supplier:  Biomatik Corporation EKU02874
Encompass_Preferred
Human Calcyclin Binding Protein (CACYBP) ELISA Kit for detecting CACYBP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.12-5000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9HB71. Gene ID: 27101. Target Synonyms: SIP; GIG5; PNAS-107; RP1-102G20.6; S100A6BP; S100A6-binding protein; Siah-interacting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9085