Learn More
Biomatik Corporation Human Calmegin (CLGN) ELISA Kit
Supplier:  Biomatik Corporation EKU12199
Encompass_Preferred
Human Calmegin (CLGN) ELISA Kit for detecting CLGN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14967. Gene ID: 1047. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8009