Learn More
Sino Biological Anti-CALML5 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11783MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CALML5 (rh CALML5; Catalog#11783-H20E; AAH39172.1; Met 1-Glu 146). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4559