Learn More
Sino Biological Anti-CALML5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11783RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CALML5 (rh CALML5; Catalog#11783-H20E; AAH39172.1; Met1-Glu146). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4555