Learn More
Sino Biological Anti-CALML5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11783RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CALML5 (rh CALML5; Catalog#11783-H20E; AAH39172.1; Met 1-Glu 146). CALML5 specific IgG was purified by Human CALML5 affinity chromatography.
Catalog No. 50-255-4554