Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CALML5 Partial ORF (NP_059118, 47 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89937865 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-865 25 μg
89-937-864 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-937-865 Supplier Abnova™ Supplier No. H00051806Q01L
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a novel calcium binding protein expressed in the epidermis and related to the calmodulin family of calcium binding proteins. Functional studies with recombinant protein demonstrate it does bind calcium and undergoes a conformational change when it does so. Abundant expression is detected only in reconstructed epidermis and is restricted to differentiating keratinocytes. In addition, it can associate with transglutaminase 3, shown to be a key enzyme in the terminal differentiation of keratinocytes. [provided by RefSeq]

Sequence: AQLRKLISEVDSDGDGEISFQEFLTAARKARAGLEDLQVAFRAFDQDGDGHITVDELRRAMAGLGQPLPQEELDAMIREADVDQDGRVNYEEFARMLAQE

Specifications

Accession Number NP_059118
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51806
Molecular Weight (g/mol) 36.74kDa
Name CALML5 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen AQLRKLISEVDSDGDGEISFQEFLTAARKARAGLEDLQVAFRAFDQDGDGHITVDELRRAMAGLGQPLPQEELDAMIREADVDQDGRVNYEEFARMLAQE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CLSP
Common Name CALML5
Gene Symbol CALML5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.