Learn More
Biomatik Corporation Human Calmodulin Like Protein 3 (CALML3) ELISA Kit
Supplier:  Biomatik Corporation EKU02879
Encompass_Preferred
Human Calmodulin Like Protein 3 (CALML3) ELISA Kit for detecting CALML3 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27482. Gene ID: 810. Target Synonyms: CLP; Calmodulin-related protein NB-1; CaM-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5461