Learn More
Bioworld HUMAN CALMODULIN (RECOMBINANT)
Supplier:  Bioworld 220602671
Encompass_Preferred
Recombinant Human Calmodulin Source: E.coli Description : Calmodulin (CaM) is an intracellular receptor protein for Ca2+-ions. It activates the myosin light chain kinase which is the catalyst of myosin phosphorylation.CaM participate...
Catalog No. 50-253-365