Learn More
Biomatik Corporation Human Calnexin (CNX) ELISA Kit
Supplier:  Biomatik Corporation EKU02881
Encompass_Preferred
Human Calnexin (CNX) ELISA Kit for detecting CNX in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27824. Gene ID: 821. Target Synonyms: CANX; CNX; IP90; P90; Major Histocompatibility Complex Class I Antigen-Binding Protein p88. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7302