Learn More
Biomatik Corporation Human Calpain 2 (CAPN2) ELISA Kit
Supplier:  Biomatik Corporation EKU02885
Encompass_Preferred
Human Calpain 2 (CAPN2) ELISA Kit for detecting CAPN2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17655. Gene ID: 824. Target Synonyms: CANPL2; CANPml; mCANP; Calpain-2 Catalytic Subunit; Calcium-activated neutral proteinase 2; Calpain M-type; Calpain large polypeptide L2; Millimolar-calpain; Calpain 2, Large Subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9082