Learn More
Biomatik Corporation Human Calpain 6 (CAPN6) ELISA Kit
Supplier:  Biomatik Corporation EKU02887
Encompass_Preferred
Human Calpain 6 (CAPN6) ELISA Kit for detecting CAPN6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y6Q1. Gene ID: 827. Target Synonyms: CAPNX; CalpM; CANPX; Calpamodulin; Calpain-like protease X-linked. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6835