Learn More
Biomatik Corporation Human Calpain, Small Subunit 1 (CAPNS1) ELISA Kit
Supplier:  Biomatik Corporation EKU02888
Encompass_Preferred
Human Calpain, Small Subunit 1 (CAPNS1) ELISA Kit for detecting CAPNS1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04632. Gene ID: 826. Target Synonyms: CANP; 30K; Calpain 4; CANPS; CAPN4; CDPS; Calcium-activated neutral proteinase small subunit; Calcium-dependent protease small subunit; Calpain regulatory subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7430