Learn More
Biomatik Corporation Human Calpastatin (CAST) ELISA Kit
Supplier:  Biomatik Corporation EKU10696
Encompass_Preferred
Human Calpastatin (CAST) ELISA Kit for detecting CAST in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20810. Gene ID: 831. Target Synonyms: BS17; Calpain inhibitor; Sperm BS-17 component. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8308