Learn More
Biomatik Corporation Human Calponin 1, Basic (CNN1) ELISA Kit
Supplier:  Biomatik Corporation EKU02890
Encompass_Preferred
Human Calponin 1, Basic (CNN1) ELISA Kit for detecting CNN1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51911. Gene ID: 1264. Target Synonyms: SMCC; Sm-Calp; Smooth Muscle Cell Calponin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9081