Learn More
Biomatik Corporation Human Calreticulin (CALR) ELISA Kit
Supplier:  Biomatik Corporation EKU10700
Encompass_Preferred
Human Calreticulin (CALR) ELISA Kit for detecting CALR in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.47-200ng/ml. Sensitivity: 0.87ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P27797. Gene ID: 811. Target Synonyms: CRT; RO; SSA; CC1qR; ERp60; HACBP; grp60; CRTC; CRP55; Calregulin; Sicca Syndrome Antigen A; Autoantigen Ro; Endoplasmic reticulum resident protein 60. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8307