Learn More
Biomatik Corporation Human Calumenin (CALU) ELISA Kit
Supplier:  Biomatik Corporation EKU02898
Encompass_Preferred
Human Calumenin (CALU) ELISA Kit for detecting CALU in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.263ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43852. Gene ID: 813. Target Synonyms: Crocalbin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9079