Learn More
Sino Biological Anti-CAMKIV Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100868T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CaM Kinase 4/CaMKIV, and purified by antigen affinity chromatography.
Catalog No. 50-240-8448