Learn More
American Research Products Inc Human cAMP response element bi
Catalog No. 5000329370
Encompass
Human cAMP response element-binding protein (CREB) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29370 Supplier American Research Products Inc Supplier No. CSBE13527H
Human cAMP response element-binding protein (CREB) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich