Learn More
Sino Biological Anti-CAND1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100869T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CAND1, and purified by antigen affinity chromatography.
Catalog No. 50-241-1028