Learn More
Biomatik Corporation Human Cannabinoid Receptor 1, Brain (CNR1) ELISA Kit
Supplier:  Biomatik Corporation EKU02902
Encompass_Preferred
Human Cannabinoid Receptor 1, Brain (CNR1) ELISA Kit for detecting CNR1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21554. Gene ID: 1268. Target Synonyms: CANN6; CB-R; CB1; CB1A; CB1K5; CNR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9078