Learn More
Biomatik Corporation Human Canopy 2 Homolog (CNPY2) ELISA Kit
Supplier:  Biomatik Corporation EKU02905
Encompass_Preferred
Human Canopy 2 Homolog (CNPY2) ELISA Kit for detecting CNPY2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2B0. Target Synonyms: ZSIG9; Cnpy2; MSAP; TMEM4; Transmembrane Protein 4; MIR-interacting saposin-like protein; Putative secreted protein Zsig9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5891