Learn More
Sino Biological Anti-CAP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107669T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CAP1, and purified by antigen affinity chromatography.
Catalog No. 50-240-9375