Learn More
Sino Biological Anti-CAPG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200979T44100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CAPG fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3959