Learn More
Sino Biological Anti-Calpain 6/CAPN6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107011T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Calpain 6/CAPN6, and purified by antigen affinity chromatography.
Catalog No. 50-240-8218