Learn More
American Research Products Inc Human Carbonic Anhydrase 3 CA
Catalog No. 5000328637
Encompass
Human Carbonic Anhydrase 3 (CA3) ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28637 Supplier American Research Products Inc Supplier No. CSBE15962H
Human Carbonic Anhydrase 3 (CA3) ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich