Learn More
Sino Biological Recombinant Human Carbonic Anhydrase IX/CA9 Protein (hFc Tag)
Supplier:  Sino Biological 10107H02H50
Encompass_Preferred
A DNA sequence encoding the human carbonic anhydrase IX (CA9) precursor (NP_001207.2) (Met 1-Asp 414) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6694