Learn More
Sino Biological Recombinant Human Carbonic Anhydrase VB Protein (His Tag)
Supplier:  Sino Biological 12285H08E50
Encompass_Preferred
A DNA sequence encoding the mature form of human CA5B (Q9Y2D0) (Cys 34-Pro 317) was fused with a polyhistidine tag at the C-terminus and an initial Met at the N-terminus.
Catalog No. 50-224-7016