Learn More
Biomatik Corporation Human Carbonic Anhydrase VB, Mitochondrial (CA5B) ELISA Kit
Supplier:  Biomatik Corporation EKU02925
Encompass_Preferred
Human Carbonic Anhydrase VB, Mitochondrial (CA5B) ELISA Kit for detecting CA5B in serum, plasma, saliva, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2D0. Gene ID: 11238. Target Synonyms: CA-VB; CAVB; Carbonate dehydratase VB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4364