Learn More
Sino Biological Recombinant Human carbonic anhydrase XII/CA12 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10617H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CA12 (NP_001209.1) (Met 1-Gln 291) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6593