Learn More
Sino Biological Human Carbonic Anhydrase XIV/CA14 Matched ELISA Antibody Pair Set 15 Plates
Supplier: Sino Biological SEK1045815
The Human Carbonic Anhydrase XIV / CA14 ELISA Pair Set is for the quantitative determination of Human Carbonic Anhydrase XIV / CA14.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Carbonic Anhydrase XIV / CA14 coated on a 96-well plate. Standards and samples are added to the wells, and any Carbonic Anhydrase XIV / CA14 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Carbonic Anhydrase XIV / CA14 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Carbonic Anhydrase XIV / CA14 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of