Learn More
Biomatik Corporation Human Carboxylesterase 2 (CES2) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10525
Encompass_Preferred
Human Carboxylesterase 2 (CES2) Wide-range ELISA Kit for detecting CES2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.245ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00748. Gene ID: 8824. Target Synonyms: CE-2; CES2A1; PCE-2; iCE; Intestine,Liver; Methylumbelliferyl-acetate deacetylase 2; Cocaine esterase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5516