Learn More
Sino Biological Recombinant Human Carboxypeptidase A Protein (His Tag)
Supplier:  Sino Biological 10504H08H50
Encompass_Preferred
A DNA sequence encoding the human CPA1 precursor (NP_001859.1) (Met 1-Tyr 419) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-7307