Learn More
Sino Biological Recombinant Human Carboxypeptidase B1 Protein (His Tag)
Supplier:  Sino Biological 10501H08H50
Encompass_Preferred
A DNA sequence encoding the human CPB1 (NP_001862.2) (Met 1-Tyr 417) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-7306