Learn More
Sino Biological Recombinant Human Carboxypeptidase E/CPE Protein (His Tag)
Supplier:  Sino Biological 10069H08H50
Encompass_Preferred
A DNA sequence encoding the human carboxypeptidase E (CPE) precursor (NP_001864.1) (Met 1-Ser 453) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7256