Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Nucleocapsid(R203K, G204R)-His Recombinant Protein
Supplier:  Sino Biological 40588V07E1100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Nucleocapsid Protein (YP_009724397.2) (Ser2-Ala419(R203K, G204R)) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-6984