Learn More
Biomatik Corporation Human Carcinoembryonic Antigen Related Cell Adhesion Molecule 3 (CEACAM3) ELISA Kit
Supplier:  Biomatik Corporation EKU08420
Encompass_Preferred
Human Carcinoembryonic Antigen Related Cell Adhesion Molecule 3 (CEACAM3) ELISA Kit for detecting CEACAM3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40198. Gene ID: 1084. Target Synonyms: CD66d; CEA; CD66-D; CGM1; W264; W282; Carcinoembryonic antigen CGM1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6472