Learn More
Biomatik Corporation Human Carcinoembryonic Antigen Related Cell Adhesion Molecule 7 (CEACAM7) ELISA Kit
Supplier:  Biomatik Corporation EKU02949
Encompass_Preferred
Human Carcinoembryonic Antigen Related Cell Adhesion Molecule 7 (CEACAM7) ELISA Kit for detecting CEACAM7 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14002. Gene ID: 1087. Target Synonyms: CGM2; CEA; Carcinoembryonic Antigen Gene Family Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9074