Learn More
Biomatik Corporation Human Carcinoembryonic Antigen Related Cell Adhesion Molecule 1 (CEACAM1) ELISA Kit
Supplier:  Biomatik Corporation EKU02947
Encompass_Preferred
Human Carcinoembryonic Antigen Related Cell Adhesion Molecule 1 (CEACAM1) ELISA Kit for detecting CEACAM1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13688. Gene ID: 634. Target Synonyms: CD66a; BGP; BGP1; BGPI; CD66-A; Biliary Glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5326