Learn More
Lifeline Cell Technology Normal Human Cardiac Fibroblasts
SDP
Supplier:  Lifeline Cell Technology FC0060
Encompass_Preferred
Normal Human Cardiac Fibroblasts provide an ideal cell system to establish serum-free human feeder layers for human embryonic stem cell cultures, or as a model to study wound healing, toxicology, or basic cell biology.
Catalog No. NC1864124
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.