Learn More
Raybiotech Inc HUMAN CARDIAC TROPONIN I ELISA
Supplier:  Raybiotech Inc ELHCTNI1
Encompass_Preferred
RayBio Human Cardiac Troponin I (cTnI) ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 100 pg/ml - 8000 pg/mL
Catalog No. 50-104-3668